Learn More
MSC SPI Plug and Pin Gage, Black Oxide Steel, 0.3780 in. Diameter, Class ZZ Minus, 1 ea.
Catalog No. 5000511050
Encompass
This Class ZZ minus plug and pin gage is a precision-manufactured inspection tool designed for checking hole sizes, setting micrometers, and verifying various measuring instruments. Featuring a nominal diameter of 0.3780 in., this steel gage provides reliable performance for high-accuracy inspection tasks.
  • Class ZZ minus tolerance of -0.0002 in.
  • Black oxide finish for enhanced durability and corrosion resistance
  • Includes a certificate of accuracy with NIST traceability
  • Constructed from steel
  • Overall length of 2 in.
  • Suitable for setting micrometers, verniers, and no-go inspections
Catalog No. 50-005-11050 Supplier MSC Supplier No. 56247273
This Class ZZ minus plug and pin gage is a precision-manufactured inspection tool designed for checking hole sizes, setting micrometers, and verifying various measuring instruments. Featuring a nominal diameter of 0.3780 in., this steel gage provides reliable performance for high-accuracy inspection tasks.
  • Class ZZ minus tolerance of -0.0002 in.
  • Black oxide finish for enhanced durability and corrosion resistance
  • Includes a certificate of accuracy with NIST traceability
  • Constructed from steel
  • Overall length of 2 in.
  • Suitable for setting micrometers, verniers, and no-go inspections