Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.4150 in. Diameter, Black Oxide, 1 ea.
Catalog No. 5000510798
Encompass
This Class ZZ Plus plug gage is designed for precise measurement and inspection applications. It features a specific diameter of 0.4150 in. to ensure accurate Go/No Go testing in metalworking and inspection environments.
  • Diameter of 0.4150 in.
  • Black oxide coating for corrosion resistance
  • Coating provides visual indication of gage wear
  • Tolerance of 0.000200 in.
Catalog No. 50-005-10798 Supplier MSC Supplier No. 84502558
This Class ZZ Plus plug gage is designed for precise measurement and inspection applications. It features a specific diameter of 0.4150 in. to ensure accurate Go/No Go testing in metalworking and inspection environments.
  • Diameter of 0.4150 in.
  • Black oxide coating for corrosion resistance
  • Coating provides visual indication of gage wear
  • Tolerance of 0.000200 in.