Learn More
MSC SPI Swiss Precision Instruments Class ZZ Minus Pin Gage, 0.372 in. Diameter, Black Oxide, 1 ea
Catalog No. 5000510776
Encompass
This SPI Class ZZ minus plug and pin gage is a high-precision measurement tool designed for accurate hole diameter verification and calibration tasks. Featuring a durable black oxide finish, this gage provides enhanced wear resistance and surface protection for reliable performance in industrial metrology applications.
  • Diameter: 0.372 in.
  • Tolerance: -0.000200 in.
  • Finish: Black oxide
  • Classification: Class ZZ minus
Catalog No. 50-005-10776 Supplier MSC Supplier No. 56247216
This SPI Class ZZ minus plug and pin gage is a high-precision measurement tool designed for accurate hole diameter verification and calibration tasks. Featuring a durable black oxide finish, this gage provides enhanced wear resistance and surface protection for reliable performance in industrial metrology applications.
  • Diameter: 0.372 in.
  • Tolerance: -0.000200 in.
  • Finish: Black oxide
  • Classification: Class ZZ minus