Learn More
MSC SPI Class ZZ Minus Plug and Pin Gage, 0.9720 in. Diameter, Black Oxide, 1 ea.
Catalog No. 5000510703
Encompass
This Class ZZ minus plug and pin gage is designed for precise diameter measurement and quality control applications. Featuring a durable black oxide finish, this gage provides a reliable solution for checking hole sizes, hole locations, and gaging slots.
  • Diameter of measurement: 0.9720 in.
  • Tolerance: 0.0002 in. (0.0002 mm)
  • Coating: Black oxide
Catalog No. 50-005-10703 Supplier MSC Supplier No. 85666295
This Class ZZ minus plug and pin gage is designed for precise diameter measurement and quality control applications. Featuring a durable black oxide finish, this gage provides a reliable solution for checking hole sizes, hole locations, and gaging slots.
  • Diameter of measurement: 0.9720 in.
  • Tolerance: 0.0002 in. (0.0002 mm)
  • Coating: Black oxide