Learn More
MSC SPI Class ZZ Plus Plug Gage, Black Oxide, 0.1820 in. Diameter, 1 ea.
Catalog No. 5000510665
Encompass
This Class ZZ Plus plug gage is designed for high-precision measurement tasks, including go inspection and the setting of micrometers, verniers, and other measuring instruments. It features a 0.1820 in. diameter and is constructed from hardened drill rod.
  • Diameter: 0.1820 in.
  • Tolerance: 0.000200 in.
  • Black oxide finish provides corrosion resistance and allows for visual identification of gage wear
  • Hardened to RC 60-62
  • Meets ASME B89.1 standards
  • Length: 2 in.
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-10665 Supplier MSC Supplier No. 84500214
This Class ZZ Plus plug gage is designed for high-precision measurement tasks, including go inspection and the setting of micrometers, verniers, and other measuring instruments. It features a 0.1820 in. diameter and is constructed from hardened drill rod.
  • Diameter: 0.1820 in.
  • Tolerance: 0.000200 in.
  • Black oxide finish provides corrosion resistance and allows for visual identification of gage wear
  • Hardened to RC 60-62
  • Meets ASME B89.1 standards
  • Length: 2 in.
  • Includes certificate of accuracy with NIST traceability