Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.4450 in. Diameter, Black Oxide, 1 ea.
Catalog No. 5000510667
Encompass
This Class ZZ plug gage is designed for precise measurement applications with a diameter of 0.4450 in. Engineered for accuracy, it features a durable finish to ensure reliability in demanding environments.
  • Diameter of 0.4450 in.
  • Tolerance of 0.000200 in.
  • Black oxide coating prevents corrosion and shows gage wear
  • Manufactured in the USA
Catalog No. 50-005-10667 Supplier MSC Supplier No. 84502855
This Class ZZ plug gage is designed for precise measurement applications with a diameter of 0.4450 in. Engineered for accuracy, it features a durable finish to ensure reliability in demanding environments.
  • Diameter of 0.4450 in.
  • Tolerance of 0.000200 in.
  • Black oxide coating prevents corrosion and shows gage wear
  • Manufactured in the USA