Learn More
MSC SPI Class ZZ Minus Plug and Pin Gage, 0.959 in. Dia, Black Oxide, 0.000200 in. Tolerance, 1 ea.
Catalog No. 5000510606
Encompass
This SPI Class ZZ minus plug and pin gage is a precision measurement tool with a diameter of 0.959 in. Designed for accuracy, it features a black oxide finish to improve wear resistance and provide a clean surface for inspection tasks.
  • Diameter of 0.959 in.
  • Tolerance of 0.000200 in.
  • Constructed with a black oxide finish for enhanced durability
Catalog No. 50-005-10606 Supplier MSC Supplier No. 85666162
This SPI Class ZZ minus plug and pin gage is a precision measurement tool with a diameter of 0.959 in. Designed for accuracy, it features a black oxide finish to improve wear resistance and provide a clean surface for inspection tasks.
  • Diameter of 0.959 in.
  • Tolerance of 0.000200 in.
  • Constructed with a black oxide finish for enhanced durability