Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.2730 in. Diameter, 1 ea.
Catalog No. 5000516164
Encompass
This Class ZZ Plus plug gage is a high-precision inspection tool designed for accurate diameter verification. Constructed from hardened drill rod and finished with black oxide, it offers enhanced wear resistance and durability for demanding metalworking environments.
  • Diameter of 0.2730 in.
  • Tolerance of 0.000200 in.
  • Hardness rated at RC 60-62
  • Overall length of 2 in.
  • Material composition of hardened drill rod
  • Black oxide coating for improved surface finish
Catalog No. 50-005-16164 Supplier MSC Supplier No. 84501121
This Class ZZ Plus plug gage is a high-precision inspection tool designed for accurate diameter verification. Constructed from hardened drill rod and finished with black oxide, it offers enhanced wear resistance and durability for demanding metalworking environments.
  • Diameter of 0.2730 in.
  • Tolerance of 0.000200 in.
  • Hardness rated at RC 60-62
  • Overall length of 2 in.
  • Material composition of hardened drill rod
  • Black oxide coating for improved surface finish