Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.3400 in. Diameter, Black Oxide, 1 ea.
Catalog No. 5000516080
Encompass
This Class ZZ Plus plug gage is designed for precise measurement applications, featuring a diameter of 0.3400 in. and a tolerance of 0.000200 in. Its robust construction ensures reliable performance for inspection tasks in metalworking and manufacturing environments.
  • Diameter of 0.3400 in.
  • Tolerance of 0.000200 in.
  • Black oxide finish for durability
Catalog No. 50-005-16080 Supplier MSC Supplier No. 84501808
This Class ZZ Plus plug gage is designed for precise measurement applications, featuring a diameter of 0.3400 in. and a tolerance of 0.000200 in. Its robust construction ensures reliable performance for inspection tasks in metalworking and manufacturing environments.
  • Diameter of 0.3400 in.
  • Tolerance of 0.000200 in.
  • Black oxide finish for durability