Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.4950 in. Diameter, Black Oxide, 1 ea.
Catalog No. 5000515980
Encompass
This Class ZZ Plus plug gage is designed for precise measurement and verification of hole diameters. Featuring a 0.4950 in. diameter, this gage provides accurate inspection for industrial metalworking applications.
  • Diameter of 0.4950 in.
  • Black oxide finish provides corrosion resistance and helps identify gage wear
  • Tolerance of 0.0002 in.
  • Made in the USA
Catalog No. 50-005-15980 Supplier MSC Supplier No. 84503358
This Class ZZ Plus plug gage is designed for precise measurement and verification of hole diameters. Featuring a 0.4950 in. diameter, this gage provides accurate inspection for industrial metalworking applications.
  • Diameter of 0.4950 in.
  • Black oxide finish provides corrosion resistance and helps identify gage wear
  • Tolerance of 0.0002 in.
  • Made in the USA