Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.3720 in. Diameter, Black Oxide, 1/Pk.
Catalog No. 5000515480
Encompass
This Class ZZ Plus plug gage is designed for precision measurement applications. The gage features a diameter of 0.3720 in. and is constructed from hardened drill rod for durability in industrial environments.
  • Features a black oxide coating to provide corrosion resistance and allow for visual detection of gage wear
  • Manufactured from hardened drill rod
  • Tolerance of 0.000200 in.
  • Diameter of 0.3720 in.
Catalog No. 50-005-15480 Supplier MSC Supplier No. 84502129
This Class ZZ Plus plug gage is designed for precision measurement applications. The gage features a diameter of 0.3720 in. and is constructed from hardened drill rod for durability in industrial environments.
  • Features a black oxide coating to provide corrosion resistance and allow for visual detection of gage wear
  • Manufactured from hardened drill rod
  • Tolerance of 0.000200 in.
  • Diameter of 0.3720 in.