Learn More
MSC SPI Pin Gage, 0.508 in., Black Oxide, Class ZZ, 1 ea.
Catalog No. 5000515481
Encompass
The Black Oxide Plus Pin Gage is a high-precision measurement tool designed for accurate gauging of hole diameters and distances. Manufactured from hardened drill rod, this gage offers reliable performance for inspection and quality control applications.
  • Diameter of 0.5080 in.
  • Class ZZ Plus designation for high precision
  • Tolerance of 0.000200 in.
  • Hardened to RC 60-62 for durability
  • Black oxide coating prevents corrosion and identifies gage wear
  • Overall length of 2 in.
  • Made in the USA
Catalog No. 50-005-15481 Supplier MSC Supplier No. 84503481
The Black Oxide Plus Pin Gage is a high-precision measurement tool designed for accurate gauging of hole diameters and distances. Manufactured from hardened drill rod, this gage offers reliable performance for inspection and quality control applications.
  • Diameter of 0.5080 in.
  • Class ZZ Plus designation for high precision
  • Tolerance of 0.000200 in.
  • Hardened to RC 60-62 for durability
  • Black oxide coating prevents corrosion and identifies gage wear
  • Overall length of 2 in.
  • Made in the USA