Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.4060 in. Diameter, Black Oxide, 1 ea.
Catalog No. 5000515260
Encompass
This class ZZ plus plug gage is designed for precise diameter measurements. Featuring a diameter of 0.4060 in., this tool is constructed from hardened drill rod to ensure durability and accuracy in various inspection applications.
  • Hardened drill rod construction with a hardness of RC 60-62
  • Black oxide coating for corrosion resistance and clear indication of gage wear
  • Overall length of 2 in.
  • Tolerance of 0.000200 in.
Catalog No. 50-005-15260 Supplier MSC Supplier No. 84502467
This class ZZ plus plug gage is designed for precise diameter measurements. Featuring a diameter of 0.4060 in., this tool is constructed from hardened drill rod to ensure durability and accuracy in various inspection applications.
  • Hardened drill rod construction with a hardness of RC 60-62
  • Black oxide coating for corrosion resistance and clear indication of gage wear
  • Overall length of 2 in.
  • Tolerance of 0.000200 in.