Learn More
MSC SPI Class ZZ Plus Plug Gage, Black Oxide, 0.7160 in. Diameter, 0.0002 in. Tolerance, 1 ea.
Catalog No. 5000515056
Encompass
This Class ZZ Plus plug gage is designed for high-precision measuring and inspection applications. Featuring a diameter of 0.7160 in., this tool is constructed from durable hardened drill rod to ensure long-lasting performance in industrial metalworking and inspection environments.
  • Features a black oxide coating to prevent corrosion and clearly indicate gage wear
  • Manufactured from hardened drill rod with a hardness of 60 to 62 RC
  • Provides a precise measurement diameter of 0.7160 in.
  • Includes a tolerance of 0.0002 in.
Catalog No. 50-005-15056 Supplier MSC Supplier No. 56240690
This Class ZZ Plus plug gage is designed for high-precision measuring and inspection applications. Featuring a diameter of 0.7160 in., this tool is constructed from durable hardened drill rod to ensure long-lasting performance in industrial metalworking and inspection environments.
  • Features a black oxide coating to prevent corrosion and clearly indicate gage wear
  • Manufactured from hardened drill rod with a hardness of 60 to 62 RC
  • Provides a precise measurement diameter of 0.7160 in.
  • Includes a tolerance of 0.0002 in.