Learn More
MSC SPI Plug and Pin Gage, Class ZZ, 0.7180 in., Black Oxide, 1 ea
Catalog No. 5000514905
Encompass
This Class ZZ minus plug and pin gage is designed for precise measurement applications. The gage features a 0.7180 in. diameter and is finished with a black oxide coating for durability.
  • Diameter of measurement: 0.7180 in.
  • Tolerance: 0.000200 in.
  • Coating: Black oxide
Catalog No. 50-005-14905 Supplier MSC Supplier No. 56250673
This Class ZZ minus plug and pin gage is designed for precise measurement applications. The gage features a 0.7180 in. diameter and is finished with a black oxide coating for durability.
  • Diameter of measurement: 0.7180 in.
  • Tolerance: 0.000200 in.
  • Coating: Black oxide