Learn More
MSC SPI Class ZZ Minus Plug Gage, Black Oxide, 0.7780 in., 1 ea.
Catalog No. 5000514908
Encompass
This Class ZZ minus plug and pin gage is a high-precision tool designed for accurate hole diameter measurement and inspection. It is constructed from hardened drill rod and features a durable black oxide coating to provide enhanced wear resistance and corrosion protection in demanding workshop environments.
  • Diameter of 0.7780 in.
  • Hardened to RC 60-62
  • Black oxide finish provides corrosion resistance and allows for easy identification of gage wear
  • Tolerance of 0.000200 in.
  • Length of 2 in.
Catalog No. 50-005-14908 Supplier MSC Supplier No. 56251275
This Class ZZ minus plug and pin gage is a high-precision tool designed for accurate hole diameter measurement and inspection. It is constructed from hardened drill rod and features a durable black oxide coating to provide enhanced wear resistance and corrosion protection in demanding workshop environments.
  • Diameter of 0.7780 in.
  • Hardened to RC 60-62
  • Black oxide finish provides corrosion resistance and allows for easy identification of gage wear
  • Tolerance of 0.000200 in.
  • Length of 2 in.