Learn More
MSC SPI Class ZZ Minus Plug Gage, 0.125 in., Black Oxide, 1 ea.
Catalog No. 5000514800
Encompass
This Class ZZ minus plug and pin gage features a 0.1250 in. diameter designed for precision measurement applications. The gage is manufactured from hardened drill rod with a black oxide finish to improve corrosion resistance and provide a visual indicator of surface wear.
  • Hardened to RC 60 to 62 for durability
  • Black oxide coating for corrosion resistance and wear indication
  • Length of 2 in.
  • Tolerance of -0.000200 in.
Catalog No. 50-005-14800 Supplier MSC Supplier No. 56244734
This Class ZZ minus plug and pin gage features a 0.1250 in. diameter designed for precision measurement applications. The gage is manufactured from hardened drill rod with a black oxide finish to improve corrosion resistance and provide a visual indicator of surface wear.
  • Hardened to RC 60 to 62 for durability
  • Black oxide coating for corrosion resistance and wear indication
  • Length of 2 in.
  • Tolerance of -0.000200 in.