Learn More
MSC SPI Class ZZ Minus Plug and Pin Gage, 0.4140 in. Diameter, Black Oxide, 1 ea.
Catalog No. 5000514762
Encompass
This Class ZZ minus plug and pin gage is designed for precise measurement applications, featuring a nominal diameter of 0.4140 in. The gage is constructed with a durable black oxide finish.
  • Nominal diameter of 0.4140 in.
  • Class ZZ tolerance rating
  • Minus tolerance of 0.000200 in.
  • Black oxide finish for added durability
Catalog No. 50-005-14762 Supplier MSC Supplier No. 56247638
This Class ZZ minus plug and pin gage is designed for precise measurement applications, featuring a nominal diameter of 0.4140 in. The gage is constructed with a durable black oxide finish.
  • Nominal diameter of 0.4140 in.
  • Class ZZ tolerance rating
  • Minus tolerance of 0.000200 in.
  • Black oxide finish for added durability