Learn More
MSC SPI Class ZZ Minus Plug Gage, Black Oxide, 0.1470 in. Diameter, 1 ea.
Catalog No. 5000514724
Encompass
This Class ZZ minus plug gage is designed for precision inspection and measurement tasks. Featuring a 0.1470 in. diameter, this gage is constructed from hardened drill rod to ensure durability and reliability in industrial environments.
  • Hardened to RC 60-62 for extended service life
  • Black oxide finish provides corrosion resistance and allows for easy visual detection of wear
  • Class ZZ minus tolerance of -0.0002 in.
  • Overall length of 2 in.
Catalog No. 50-005-14724 Supplier MSC Supplier No. 56244957
This Class ZZ minus plug gage is designed for precision inspection and measurement tasks. Featuring a 0.1470 in. diameter, this gage is constructed from hardened drill rod to ensure durability and reliability in industrial environments.
  • Hardened to RC 60-62 for extended service life
  • Black oxide finish provides corrosion resistance and allows for easy visual detection of wear
  • Class ZZ minus tolerance of -0.0002 in.
  • Overall length of 2 in.