Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.8050 in. Diameter, 1 ea.
Catalog No. 5000514713
Encompass
This Class ZZ Plus plug gage is designed for precise measurement and inspection applications. Featuring a 0.8050 in. diameter, it provides accurate assessment for various metalworking and quality control tasks.
  • Diameter of 0.8050 in.
  • Tolerance of +0.000200 in.
  • Black oxide finish
Catalog No. 50-005-14713 Supplier MSC Supplier No. 56241607
This Class ZZ Plus plug gage is designed for precise measurement and inspection applications. Featuring a 0.8050 in. diameter, it provides accurate assessment for various metalworking and quality control tasks.
  • Diameter of 0.8050 in.
  • Tolerance of +0.000200 in.
  • Black oxide finish