Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.4360 in. Diameter, Black Oxide, 1 ea.
Catalog No. 5000514559
Encompass
This Class ZZ Plus plug gage is designed for precision measurement applications. Featuring a nominal diameter of 0.4360 in., this tool is constructed for reliable inspection tasks in metalworking and manufacturing environments.
  • Diameter of 0.4360 in.
  • Tolerance of 0.0002
  • Hardened drill rod construction with a hardness of RC 60 to 62
  • Black oxide coating prevents corrosion and helps identify gage wear
  • Overall length of 2 in.
Catalog No. 50-005-14559 Supplier MSC Supplier No. 84502764
This Class ZZ Plus plug gage is designed for precision measurement applications. Featuring a nominal diameter of 0.4360 in., this tool is constructed for reliable inspection tasks in metalworking and manufacturing environments.
  • Diameter of 0.4360 in.
  • Tolerance of 0.0002
  • Hardened drill rod construction with a hardness of RC 60 to 62
  • Black oxide coating prevents corrosion and helps identify gage wear
  • Overall length of 2 in.