Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC SPI Caliper Accessory Kit, 9 Piece, for 4-8 in Calipers

Catalog No. 5000511329
Encompass

This accessory kit provides expanded functionality for calipers. It is designed for use with 4, 6, and 8 inch dial, vernier, and electronic calipers, offering various contact points and an Allen wrench to adapt instruments for different measurement applications.

  • Includes two ball end points
  • Includes two conical contact points
  • Includes two flat contact points
  • Includes two stem points
  • Allen wrench included
  • Instructions provided
  • Contains 9 pieces
  • For use with 4, 6 & 8 inch dial, vernier & electronic calipers

Catalog No. 50-005-11329 Supplier MSC Supplier No. 06410120
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This accessory kit provides expanded functionality for calipers. It is designed for use with 4, 6, and 8 inch dial, vernier, and electronic calipers, offering various contact points and an Allen wrench to adapt instruments for different measurement applications.

  • Includes two ball end points
  • Includes two conical contact points
  • Includes two flat contact points
  • Includes two stem points
  • Allen wrench included
  • Instructions provided
  • Contains 9 pieces
  • For use with 4, 6 & 8 inch dial, vernier & electronic calipers

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.