Learn More
MSC SPI Caliper Attachment, Groovemaster, 7.5 mm Min. Bore, Hard Chrome Plated
Catalog No. 5000513539
Encompass
This caliper attachment is designed for quick and accurate measurements. It is used to measure the internal and external diameters of retaining ring grooves, o-ring grooves, and undercuts. It is precision machined from steel and hard chrome plated for wear resistance. This Groovemaster fits all 6 in vernier, dial, and digital calipers with a maximum jaw thickness of 0.140 in. It can measure internal grooves from 0.200 in up to the capacity of the caliper.
  • Highly effective for quick and accurate measurements of internal and external diameters of retaining ring grooves, o-ring grooves and undercuts.
  • Available individually and in convenient sets.
  • Precision machined from steel, hard chrome plated for extra wear resistance.
  • Fits all 6 in vernier, dial and digital calipers with a maximum jaw-thickness of 0.140 in.
  • Measures internal grooves from 0.200 in up to the capacity of the caliper.
Catalog No. 50-005-13539 Supplier MSC Supplier No. 01541697
This caliper attachment is designed for quick and accurate measurements. It is used to measure the internal and external diameters of retaining ring grooves, o-ring grooves, and undercuts. It is precision machined from steel and hard chrome plated for wear resistance. This Groovemaster fits all 6 in vernier, dial, and digital calipers with a maximum jaw thickness of 0.140 in. It can measure internal grooves from 0.200 in up to the capacity of the caliper.
  • Highly effective for quick and accurate measurements of internal and external diameters of retaining ring grooves, o-ring grooves and undercuts.
  • Available individually and in convenient sets.
  • Precision machined from steel, hard chrome plated for extra wear resistance.
  • Fits all 6 in vernier, dial and digital calipers with a maximum jaw-thickness of 0.140 in.
  • Measures internal grooves from 0.200 in up to the capacity of the caliper.