Learn More
MSC SPI Caliper Set, 0.0005 in Resolution, 0-8 in Range, Stainless Steel
Catalog No. 5000530397
Encompass
This caliper set provides precision measurements for engineering and scientific applications. Featuring a 0.0005 in resolution and a 0-8 in measuring range, these calipers are constructed from durable stainless steel for longevity and corrosion resistance.
  • Resolution of 0.0005 in.
  • Measuring range of 0-8 in.
  • Durable stainless steel construction for caliper body and jaws.
  • Suitable for precision measurements in engineering and scientific applications.
  • Includes a 9 in fluorescent clock.
  • Comes with a protective case for storage.
Catalog No. 50-005-30397 Supplier MSC Supplier No. 99611964
This caliper set provides precision measurements for engineering and scientific applications. Featuring a 0.0005 in resolution and a 0-8 in measuring range, these calipers are constructed from durable stainless steel for longevity and corrosion resistance.
  • Resolution of 0.0005 in.
  • Measuring range of 0-8 in.
  • Durable stainless steel construction for caliper body and jaws.
  • Suitable for precision measurements in engineering and scientific applications.
  • Includes a 9 in fluorescent clock.
  • Comes with a protective case for storage.