Learn More
MSC SPI Center Finder Set, 0.0002 inch Accuracy, 0.1 inch Head, 5 Piece
Catalog No. 5000529784
Encompass
This mechanical center finder set features a single end with a 0.1000 inch head diameter and an accuracy of 0.0002 " The 5-piece set includes a holder, four attachments, and a case.
  • Mechanical center finder set
  • Single end type
  • 0.1000 inch head diameter
  • 0.0002 inch accuracy
  • Includes holder, four attachments, and case
Catalog No. 50-005-29784 Supplier MSC Supplier No. 01767573
This mechanical center finder set features a single end with a 0.1000 inch head diameter and an accuracy of 0.0002 " The 5-piece set includes a holder, four attachments, and a case.
  • Mechanical center finder set
  • Single end type
  • 0.1000 inch head diameter
  • 0.0002 inch accuracy
  • Includes holder, four attachments, and case