Learn More
MSC SPI Clamp, For Dovetail Test Indicators, 2 in Shank
Catalog No. 5000536272
Encompass
This clamp is designed for use with dovetail style dial test indicators. It features a 2 inch shank length and a 3 inch overall length, making it suitable for various precision measurement applications.
  • Designed for dovetail style dial test indicators.
  • Features a 2 inch shank length.
  • Has a 3 inch overall length.
Catalog No. 50-005-36272 Supplier MSC Supplier No. 08655516
This clamp is designed for use with dovetail style dial test indicators. It features a 2 inch shank length and a 3 inch overall length, making it suitable for various precision measurement applications.
  • Designed for dovetail style dial test indicators.
  • Features a 2 inch shank length.
  • Has a 3 inch overall length.