Learn More
MSC SPI Class ZZ Minus Gage, 0.6670 in, Steel
Catalog No. 5000528813
Encompass
This high-precision Class ZZ Minus plug and pin gage is designed for accurate measurement. It features a specific diameter of 0.6670 in and is constructed from durable steel, making it suitable for various industrial and laboratory applications.
  • Diameter of measurement: 0.6670 in
  • Tolerance: 0.000200 in
  • Material: Steel
  • System of measurement: Decimal inch
Catalog No. 50-005-28813 Supplier MSC Supplier No. 78378635
This high-precision Class ZZ Minus plug and pin gage is designed for accurate measurement. It features a specific diameter of 0.6670 in and is constructed from durable steel, making it suitable for various industrial and laboratory applications.
  • Diameter of measurement: 0.6670 in
  • Tolerance: 0.000200 in
  • Material: Steel
  • System of measurement: Decimal inch