Learn More
MSC SPI Class ZZ Minus Pin Gage, 0.091 in Diameter, Black Oxide
Catalog No. 5000528804
Encompass
The SPI Class ZZ Minus Pin Gage is designed for precise measurement and inspection applications. It features a specific diameter for accurate gaging and a black oxide coating that helps prevent corrosion and visibly indicates wear, ensuring its durability and reliability in various industrial settings.
  • Black oxide coating prevents corrosion and shows gage wear
  • Made from hardened drill rod
  • Hardness RC 60-62
  • Used for inspecting measurements
Catalog No. 50-005-28804 Supplier MSC Supplier No. 56244395
The SPI Class ZZ Minus Pin Gage is designed for precise measurement and inspection applications. It features a specific diameter for accurate gaging and a black oxide coating that helps prevent corrosion and visibly indicates wear, ensuring its durability and reliability in various industrial settings.
  • Black oxide coating prevents corrosion and shows gage wear
  • Made from hardened drill rod
  • Hardness RC 60-62
  • Used for inspecting measurements