Learn More
MSC SPI Class ZZ Minus Plug Gage, 0.9580 in, 0.0002 in
Catalog No. 5000531419
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection tasks. It features a specific diameter and tight tolerance for accuracy, and a black oxide finish for durability.
  • Diameter of 0.9580 inch
  • Tolerance of 0.0002 inch
  • Black oxide coating for durability
Catalog No. 50-005-31419 Supplier MSC Supplier No. 85666154
Add to Cart
Add to Cart
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection tasks. It features a specific diameter and tight tolerance for accuracy, and a black oxide finish for durability.
  • Diameter of 0.9580 inch
  • Tolerance of 0.0002 inch
  • Black oxide coating for durability