Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.0410 in Dia, Black Oxide
Catalog No. 5000515061
Encompass
The SPI Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. This gage features a 0.0410 inch diameter and a durable black oxide finish, making it suitable for checking bore sizes and other critical dimensions with high accuracy.
  • Class ZZ Minus Plug & Pin Gage
  • Diameter of measurement is 0.0410 inch
  • Tolerance of -0.000200 inch
  • Black oxide coating for durability
Catalog No. 50-005-15061 Supplier MSC Supplier No. 56243884
The SPI Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. This gage features a 0.0410 inch diameter and a durable black oxide finish, making it suitable for checking bore sizes and other critical dimensions with high accuracy.
  • Class ZZ Minus Plug & Pin Gage
  • Diameter of measurement is 0.0410 inch
  • Tolerance of -0.000200 inch
  • Black oxide coating for durability