Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.0450 in Diameter, Black Oxide, -0.000200 in Tolerance
Catalog No. 5000531130
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a specific diameter and a durable black oxide coating, making it suitable for environments requiring exact dimensional verification. The gage provides a precise tolerance for accurate readings.
  • Diameter of measurement: 0.0450 in
  • Tolerance: -0.000200 in
  • Coating/finish: Black oxide
  • Country of origin: China
Catalog No. 50-005-31130 Supplier MSC Supplier No. 56243934
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a specific diameter and a durable black oxide coating, making it suitable for environments requiring exact dimensional verification. The gage provides a precise tolerance for accurate readings.
  • Diameter of measurement: 0.0450 in
  • Tolerance: -0.000200 in
  • Coating/finish: Black oxide
  • Country of origin: China