Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.0600 in Diameter, Black Oxide
Catalog No. 5000528871
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement of small diameters. With a nominal diameter of 0.0600 inches, it is manufactured to exact specifications, ensuring accuracy for quality control and inspection tasks. The black oxide finish provides durability and corrosion resistance.
  • Precisely measures diameters up to 0.0600 inches
  • Features a tight tolerance of -0.000200 inches
  • Enhanced durability with a black oxide finish
Catalog No. 50-005-28871 Supplier MSC Supplier No. 56244080
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement of small diameters. With a nominal diameter of 0.0600 inches, it is manufactured to exact specifications, ensuring accuracy for quality control and inspection tasks. The black oxide finish provides durability and corrosion resistance.
  • Precisely measures diameters up to 0.0600 inches
  • Features a tight tolerance of -0.000200 inches
  • Enhanced durability with a black oxide finish