Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.0610 in Diameter, Steel, 0.0002 in Tolerance
Catalog No. 5000530095
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement and inspection tasks, such as setting micrometers and verifying other measuring devices. It provides accurate dimensions for quality control and calibration.
  • 0.0610 in diameter
  • 0.0002 in tolerance
  • Made of steel
  • Suitable for no-go inspection
  • Ideal for verifying verniers and other measuring devices
Catalog No. 50-005-30095 Supplier MSC Supplier No. 78372570
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement and inspection tasks, such as setting micrometers and verifying other measuring devices. It provides accurate dimensions for quality control and calibration.
  • 0.0610 in diameter
  • 0.0002 in tolerance
  • Made of steel
  • Suitable for no-go inspection
  • Ideal for verifying verniers and other measuring devices