Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.0870 in Dia, Black Oxide
Catalog No. 5000531131
Encompass
This Class ZZ Minus Plug & Pin Gage is precisely manufactured for accurate measurement. Featuring a 0.0870 inch diameter and a durable black oxide finish, it is designed for demanding industrial applications requiring high precision.
  • Precisely manufactured for accurate measurements.
  • Features a diameter of 0.0870 "
  • Engineered with a tolerance of -0.000200 "
  • Coated with black oxide for increased durability and corrosion resistance.
Catalog No. 50-005-31131 Supplier MSC Supplier No. 56244353
This Class ZZ Minus Plug & Pin Gage is precisely manufactured for accurate measurement. Featuring a 0.0870 inch diameter and a durable black oxide finish, it is designed for demanding industrial applications requiring high precision.
  • Precisely manufactured for accurate measurements.
  • Features a diameter of 0.0870 "
  • Engineered with a tolerance of -0.000200 "
  • Coated with black oxide for increased durability and corrosion resistance.