Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.0940 in Dia, -0.000200 in Tolerance
Catalog No. 5000527781
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for accurate dimensional inspection, offering precise gaging capabilities. It is suitable for applications requiring close tolerance measurements and features a protective finish.
  • Measures with a diameter of 0.0940 inches.
  • Offers a precise tolerance of -0.000200 inches (-0.0002 mm).
  • Features a black oxide coating.
  • Originates from China.
Catalog No. 50-005-27781 Supplier MSC Supplier No. 56244429
This Class ZZ Minus Plug & Pin Gage is designed for accurate dimensional inspection, offering precise gaging capabilities. It is suitable for applications requiring close tolerance measurements and features a protective finish.
  • Measures with a diameter of 0.0940 inches.
  • Offers a precise tolerance of -0.000200 inches (-0.0002 mm).
  • Features a black oxide coating.
  • Originates from China.