Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.0980" Diameter, Black Oxide
Catalog No. 5000528801
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a 0.0980 inch diameter and a durable black oxide finish.
  • Measures 0.0980 inch in diameter
  • Features a tolerance of -0.000200 inch
  • Equipped with a black oxide finish
  • Manufactured in China
Catalog No. 50-005-28801 Supplier MSC Supplier No. 56244460
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a 0.0980 inch diameter and a durable black oxide finish.
  • Measures 0.0980 inch in diameter
  • Features a tolerance of -0.000200 inch
  • Equipped with a black oxide finish
  • Manufactured in China