Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.1010 in Dia, -0.000200 in Tolerance, Black Oxide
Catalog No. 5000530695
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement with a 0.1010 inch diameter and a -0.000200 inch tolerance. It features a durable black oxide finish, ensuring reliable performance for various applications.
  • Product can be exchanged or refunded if it doesn't meet expectations
  • Return within 30 days of shipment in its original packaging (excluding hazardous materials and cut-to-length products)
Catalog No. 50-005-30695 Supplier MSC Supplier No. 56244494
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement with a 0.1010 inch diameter and a -0.000200 inch tolerance. It features a durable black oxide finish, ensuring reliable performance for various applications.
  • Product can be exchanged or refunded if it doesn't meet expectations
  • Return within 30 days of shipment in its original packaging (excluding hazardous materials and cut-to-length products)