Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.1300 in Diameter, Steel
Catalog No. 5000530489
Encompass
This Class ZZ Minus Plug & Pin Gage is precision-engineered for inspection and measurement tasks. Constructed from durable steel, it features a diameter of 0.1300 inches, adhering to Class ZZ Minus tolerance standards for accuracy.
  • Designed for precise inspection and measurement applications.
  • Manufactured from steel for enhanced durability and longevity.
  • Features a 0.1300 inch diameter for specific gauging requirements.
  • Adheres to Class ZZ Minus tolerance standards, ensuring high accuracy with a tolerance of 0.000200 inches.
Catalog No. 50-005-30489 Supplier MSC Supplier No. 78373263
This Class ZZ Minus Plug & Pin Gage is precision-engineered for inspection and measurement tasks. Constructed from durable steel, it features a diameter of 0.1300 inches, adhering to Class ZZ Minus tolerance standards for accuracy.
  • Designed for precise inspection and measurement applications.
  • Manufactured from steel for enhanced durability and longevity.
  • Features a 0.1300 inch diameter for specific gauging requirements.
  • Adheres to Class ZZ Minus tolerance standards, ensuring high accuracy with a tolerance of 0.000200 inches.