Learn More
MSC SPI Class ZZ Minus Plug and Pin Gage, 0.1440 in Diameter, Steel
Catalog No. 5000528977
Encompass
This product is a Class ZZ Minus Plug and Pin Gage, essential for precise measurement and inspection. It features a 0.1440 inch diameter and is constructed from durable steel, ensuring accuracy and longevity in various industrial applications.
  • Class ZZ minus plug and pin gage
  • 0.1440 inch diameter for accurate measurements
  • Tolerance of 0.000200 inch (0.0002 mm)
  • Constructed from steel for durability
  • Designed for decimal inch measurement systems
Catalog No. 50-005-28977 Supplier MSC Supplier No. 78373404
Add to Cart
Add to Cart
This product is a Class ZZ Minus Plug and Pin Gage, essential for precise measurement and inspection. It features a 0.1440 inch diameter and is constructed from durable steel, ensuring accuracy and longevity in various industrial applications.
  • Class ZZ minus plug and pin gage
  • 0.1440 inch diameter for accurate measurements
  • Tolerance of 0.000200 inch (0.0002 mm)
  • Constructed from steel for durability
  • Designed for decimal inch measurement systems