Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.1630 in, Steel
Catalog No. 5000528692
Encompass
This is a Class ZZ Minus plug and pin gage designed for precise measurement applications. Manufactured from durable steel, it features a diameter of 0.1630 inches, providing accurate and reliable gauging for various industrial and scientific needs.
  • Precise diameter of 0.1630 inches.
  • Class ZZ Minus for specific gauging requirements.
  • Constructed from durable steel.
  • High tolerance of 0.000200 inches for accurate results.
Catalog No. 50-005-28692 Supplier MSC Supplier No. 78373594
This is a Class ZZ Minus plug and pin gage designed for precise measurement applications. Manufactured from durable steel, it features a diameter of 0.1630 inches, providing accurate and reliable gauging for various industrial and scientific needs.
  • Precise diameter of 0.1630 inches.
  • Class ZZ Minus for specific gauging requirements.
  • Constructed from durable steel.
  • High tolerance of 0.000200 inches for accurate results.