Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.1660 in Diameter, Black Oxide
Catalog No. 5000529051
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a specific diameter and a durable finish, making it suitable for various quality control and inspection tasks.
  • Class ZZ Minus Plug & Pin Gage
  • Diameter of 0.1660 in
  • Black oxide finish
  • Tolerance of -0.000200 in
Catalog No. 50-005-29051 Supplier MSC Supplier No. 56245145
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a specific diameter and a durable finish, making it suitable for various quality control and inspection tasks.
  • Class ZZ Minus Plug & Pin Gage
  • Diameter of 0.1660 in
  • Black oxide finish
  • Tolerance of -0.000200 in