Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.1860 in Diameter, Steel
Catalog No. 5000528971
Encompass
The Class ZZ Minus Plug & Pin Gage is a precise measuring tool designed for accurate diameter checks. It features a diameter of 0.1860 inches and is constructed from durable steel, ensuring reliability and longevity for various applications. With a tight tolerance of 0.000200 inches, this gage provides consistent and dependable measurements.
  • Has a diameter of 0.1860 inches.
  • Constructed from steel.
  • Offers a tolerance of 0.000200 inches.
Catalog No. 50-005-28971 Supplier MSC Supplier No. 78373826
The Class ZZ Minus Plug & Pin Gage is a precise measuring tool designed for accurate diameter checks. It features a diameter of 0.1860 inches and is constructed from durable steel, ensuring reliability and longevity for various applications. With a tight tolerance of 0.000200 inches, this gage provides consistent and dependable measurements.
  • Has a diameter of 0.1860 inches.
  • Constructed from steel.
  • Offers a tolerance of 0.000200 inches.