Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.2230 in Diameter, -0.000200 in Tolerance, Black Oxide
Catalog No. 5000527503
Encompass
This plug and pin gage features a precise measurement diameter of 0.2230 in with a tight tolerance of -0.000200 in (-0.0002 mm). The gage is finished with a black oxide coating, providing durability, and is suitable for various measuring and inspecting applications.
  • Precise measurement diameter of 0.2230 in
  • Tight tolerance of -0.000200 in (-0.0002 mm)
  • Black oxide coating for enhanced durability
  • Suitable for various measuring and inspecting applications
Catalog No. 50-005-27503 Supplier MSC Supplier No. 56245723
This plug and pin gage features a precise measurement diameter of 0.2230 in with a tight tolerance of -0.000200 in (-0.0002 mm). The gage is finished with a black oxide coating, providing durability, and is suitable for various measuring and inspecting applications.
  • Precise measurement diameter of 0.2230 in
  • Tight tolerance of -0.000200 in (-0.0002 mm)
  • Black oxide coating for enhanced durability
  • Suitable for various measuring and inspecting applications