Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.3620 in, Black Oxide
Catalog No. 5000531978
Encompass
SPI Class ZZ Minus Plug & Pin Gages are precision tools designed for accurately measuring bore and hole diameters. This gage is engineered for high accuracy and durability, featuring a specific diameter and protective coating.
  • Measures 0.3620 inch diameter
  • Features a tolerance of -0.000200 inch
  • Coated with black oxide for enhanced durability
  • Manufactured in China
Catalog No. 50-005-31978 Supplier MSC Supplier No. 56247117
SPI Class ZZ Minus Plug & Pin Gages are precision tools designed for accurately measuring bore and hole diameters. This gage is engineered for high accuracy and durability, featuring a specific diameter and protective coating.
  • Measures 0.3620 inch diameter
  • Features a tolerance of -0.000200 inch
  • Coated with black oxide for enhanced durability
  • Manufactured in China