Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.3760 inch Dia, Black Oxide
Catalog No. 5000527863
Encompass
This plug and pin gage is designed for precise measurement applications. It features a Class ZZ Minus tolerance and a black oxide finish for durability.
  • Precise measurement with a 0.3760 inch diameter
  • Features a Class ZZ Minus tolerance
  • Durable black oxide coating
  • Country of origin is China
Catalog No. 50-005-27863 Supplier MSC Supplier No. 56247257
This plug and pin gage is designed for precise measurement applications. It features a Class ZZ Minus tolerance and a black oxide finish for durability.
  • Precise measurement with a 0.3760 inch diameter
  • Features a Class ZZ Minus tolerance
  • Durable black oxide coating
  • Country of origin is China