Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.4260 in, 0.000200 in Tolerance, Steel
Catalog No. 5000531044
Encompass
This plug and pin gage is designed for precise measurement and verification. It features a specific diameter and tight tolerance, making it suitable for various quality control and inspection applications requiring high accuracy.
  • Diameter of measurement: 0.4260 in
  • Tolerance: 0.000200 in
  • Material: Steel
Catalog No. 50-005-31044 Supplier MSC Supplier No. 78376225
This plug and pin gage is designed for precise measurement and verification. It features a specific diameter and tight tolerance, making it suitable for various quality control and inspection applications requiring high accuracy.
  • Diameter of measurement: 0.4260 in
  • Tolerance: 0.000200 in
  • Material: Steel