Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.4810 in Diameter, Black Oxide
Catalog No. 5000528775
Encompass
This Class ZZ minus plug and pin gage features a 0.4810 in diameter with a tolerance of 0.000200 in, and is finished with black oxide for enhanced durability.
  • Class ZZ minus plug and pin gage.
  • 0.4810 in diameter for precise measurements.
  • Tolerance of 0.000200 in ensures accuracy.
  • Black oxide finish.
  • Manufactured in China.
Catalog No. 50-005-28775 Supplier MSC Supplier No. 56248305
This Class ZZ minus plug and pin gage features a 0.4810 in diameter with a tolerance of 0.000200 in, and is finished with black oxide for enhanced durability.
  • Class ZZ minus plug and pin gage.
  • 0.4810 in diameter for precise measurements.
  • Tolerance of 0.000200 in ensures accuracy.
  • Black oxide finish.
  • Manufactured in China.