Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.4970 in, Steel
Catalog No. 5000530094
Encompass
The SPI Class ZZ Minus Plug & Pin Gage is a precision measurement tool designed for accurate dimensional inspection. It features a 0.4970 inch diameter and is made from durable steel, ensuring reliable performance for various industrial and laboratory applications.
  • Designed for precise dimensional inspection
  • Features a 0.4970 inch diameter
  • Offers a tight tolerance of 0.000200 inch
  • Constructed from durable steel
  • Manufactured to Class ZZ minus standards
Catalog No. 50-005-30094 Supplier MSC Supplier No. 78376936
The SPI Class ZZ Minus Plug & Pin Gage is a precision measurement tool designed for accurate dimensional inspection. It features a 0.4970 inch diameter and is made from durable steel, ensuring reliable performance for various industrial and laboratory applications.
  • Designed for precise dimensional inspection
  • Features a 0.4970 inch diameter
  • Offers a tight tolerance of 0.000200 inch
  • Constructed from durable steel
  • Manufactured to Class ZZ minus standards