Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.5010 in Dia, Black Oxide
Catalog No. 5000527935
Encompass
The product is a Class ZZ Minus Plug & Pin Gage, designed for precise measurement. It features a 0.5010 inch diameter and a durable black oxide finish.
  • Diameter of measurement is 0.5010 in
  • Tolerance is 0.000200 in
  • Features a black oxide finish
Catalog No. 50-005-27935 Supplier MSC Supplier No. 56248503
Add to Cart
Add to Cart
The product is a Class ZZ Minus Plug & Pin Gage, designed for precise measurement. It features a 0.5010 inch diameter and a durable black oxide finish.
  • Diameter of measurement is 0.5010 in
  • Tolerance is 0.000200 in
  • Features a black oxide finish