Learn More
MSC SPI Class ZZ Minus Plug & Pin Gage, 0.5080 in Diameter, Black Oxide
Catalog No. 5000531741
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications, offering a high degree of accuracy. It features a durable black oxide coating and a specific diameter for consistent and reliable gauging.
  • Designed for precise measurement applications
  • Features a 0.5080 inch diameter
  • Maintains a tight tolerance of 0.000200 inch
  • Coated with black oxide for durability and corrosion resistance
  • Suitable for various gauging needs
Catalog No. 50-005-31741 Supplier MSC Supplier No. 56248578
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications, offering a high degree of accuracy. It features a durable black oxide coating and a specific diameter for consistent and reliable gauging.
  • Designed for precise measurement applications
  • Features a 0.5080 inch diameter
  • Maintains a tight tolerance of 0.000200 inch
  • Coated with black oxide for durability and corrosion resistance
  • Suitable for various gauging needs